missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopentylpropionyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11403308
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-97-2
Molecular Formula C8H13ClO
Molecular Weight (g/mol) 160.64
MDL Number MFCD00001391
InChI Key SZQVEGOXJYTLLB-UHFFFAOYSA-N
Synonym 3-cyclopentylpropionyl chloride, cyclopentylpropionyl chloride, cyclopentanepropionyl chloride, 3-cyclopentyl propionyl chloride, cyclopentanepropanoyl-chloride, pubchem21363, cyclopentylpropionylchloride, cyclopentanepropanoylchloride, 3-cyclopentylpropanyl chloride, cyclopentane propionyl chloride
PubChem CID 66039
IUPAC Name 3-cyclopentylpropanoyl chloride
SMILES ClC(=O)CCC1CCCC1

Specifications

Density 1.049
Boiling Point 199°C to 200°C
Flash Point 84°C (184°F)
Odor Acrid
Refractive Index 1.4645
Quantity 100 g
UN Number UN3265
Beilstein 1856952
Sensitivity Moisture sensitive
Formula Weight 160.64
Percent Purity 98%
Chemical Name or Material 3-Cyclopentylpropionyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.