missing translation for 'onlineSavingsMsg'
Learn More

(3-Cyanopropyl)trichlorosilane, 98%, Thermo Scientific Chemicals

Product Code. 11409060
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-27-8
Molecular Formula C4H6Cl3NSi
Molecular Weight (g/mol) 202.534
MDL Number MFCD00013832
InChI Key HMFFOEBLYHLRQN-UHFFFAOYSA-N
Synonym 3-cyanopropyltrichlorosilane, butanenitrile, 4-trichlorosilyl, 4-trichlorosilyl butyronitrile, 4-cyanobutyltrichlorosilane, butyronitrile, 4-trichlorosilyl, trichloro 3-cyanopropyl silane, trichlor-3-kyanpropylsilan, silane, trichloro 3-cyanopropyl, unii-05m8iu15r8, gamma-cyanopropyltrichlorosilane
PubChem CID 14063
IUPAC Name 4-trichlorosilylbutanenitrile
SMILES C(CC#N)C[Si](Cl)(Cl)Cl

Specifications

Boiling Point 237°C to 238°C
Quantity 100 g
Linear Formula Cl3Si(CH2)3CN
UN Number UN2987
Beilstein 906924
Flash Point 92°C (198°F)
Formula Weight 202.54
Percent Purity 98%
Odor Pungent
Refractive Index 1.463
Sensitivity Moisture sensitive
Density 1.28
Chemical Name or Material (3-Cyanopropyl)trichlorosilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'