missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-2-methoxypyridine, 97+%, Thermo Scientific™

54.87 € - 605.00 €

Chemical Identifiers

CAS 7254-34-4
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.14
MDL Number MFCD07363804
InChI Key IHWZCGMZILLKFD-UHFFFAOYSA-N
PubChem CID 252099
IUPAC Name 2-methoxypyridine-3-carbonitrile
SMILES COC1=NC=CC=C1C#N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15429067
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H52373.MD
250 mg
54.87 €
250mg
Please to purchase this item. Need a web account? Register with us today!
15409067
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H52373.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15419067
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H52373.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Cyano-2-methoxypyridine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Chemical Identifiers

7254-34-4
134.14
IHWZCGMZILLKFD-UHFFFAOYSA-N
2-methoxypyridine-3-carbonitrile
C7H6N2O
MFCD07363804
252099
COC1=NC=CC=C1C#N

Specifications

7254-34-4
MFCD07363804
Slightly soluble in water.
COC1=NC=CC=C1C#N
134.14
134.14
3-Cyano-2-methoxypyridine
C7H6N2O
2-methoxynicotinonitrile, 3-cyano-2-methoxypyridine, 2-methoxy-nicotinonitrile, 2-methoxy-3-cyanopyridine, 3-pyridinecarbonitrile, 2-methoxy, 2-methoxy-3-pyridinecarbonitrile, 2-methoxynicotinonitril, acmc-1bcy7, 3-pyridinecarbonitrile,2-methoxy
IHWZCGMZILLKFD-UHFFFAOYSA-N
2-methoxypyridine-3-carbonitrile
252099
≥97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications