missing translation for 'onlineSavingsMsg'
Learn More

3-Coumaranone, 97%, Thermo Scientific Chemicals

47.54 € - 661.29 €

Chemical Identifiers

CAS 7169-34-8
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.13
MDL Number MFCD00051810
InChI Key MGKPCLNUSDGXGT-UHFFFAOYSA-N
Synonym benzofuran-3 2h-one, 3-coumaranone, 3 2h-benzofuranone, 1-benzofuran-3 2h-one, coumaranone, benzofuran-3-one, coumaran-3-one, 2,3-dihydrobenzo b furan-3-one, 2,3-dihydro-1-benzofuran-3-one, 2h-benzofuran-3-one
PubChem CID 23556
IUPAC Name 1-benzofuran-3-one
SMILES O=C1COC2=CC=CC=C12
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11409522
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10202.03
1 g
47.54 €
1g
Please to purchase this item. Need a web account? Register with us today!
11419522
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10202.06
5 g
172.86 €
5g
Please to purchase this item. Need a web account? Register with us today!
11429522
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10202.14
25 g
661.29 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

3-Coumaranone is used in the aurones, [2-benzylidenebenzofuran-3(2H)-ones] by reacting with appropriate aldehyde. It is involved in the condensation reaction with aldehydes in the presence of morpholine acetate to prepare substituted 2-(arylidene)benzofuran-3(ZH)-ones. In Horner-Wadsworth-Emmons reaction, it reacts with diethyl¬ cyanomethyl¬phosphonate to give 3-(cyanomethyl)benzofurans, which is an intermediate in the synthesis of antihyperglycemic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Coumaranone is used in the aurones, [2-benzylidenebenzofuran-3(2H)-ones] by reacting with appropriate aldehyde. It is involved in the condensation reaction with aldehydes in the presence of morpholine acetate to prepare substituted 2-(arylidene)benzofuran-3(ZH)-ones. In Horner-Wadsworth-Emmons reaction, it reacts with diethyl¬ cyanomethyl¬phosphonate to give 3-(cyanomethyl)benzofurans, which is an intermediate in the synthesis of antihyperglycemic agents.

Solubility
Slightly miscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

7169-34-8
134.13
MGKPCLNUSDGXGT-UHFFFAOYSA-N
23556
O=C1COC2=CC=CC=C12
C8H6O2
MFCD00051810
benzofuran-3 2h-one, 3-coumaranone, 3 2h-benzofuranone, 1-benzofuran-3 2h-one, coumaranone, benzofuran-3-one, coumaran-3-one, 2,3-dihydrobenzo b furan-3-one, 2,3-dihydro-1-benzofuran-3-one, 2h-benzofuran-3-one
1-benzofuran-3-one

Specifications

7169-34-8
C8H6O2
115296
benzofuran-3 2h-one, 3-coumaranone, 3 2h-benzofuranone, 1-benzofuran-3 2h-one, coumaranone, benzofuran-3-one, coumaran-3-one, 2,3-dihydrobenzo b furan-3-one, 2,3-dihydro-1-benzofuran-3-one, 2h-benzofuran-3-one
MGKPCLNUSDGXGT-UHFFFAOYSA-N
1-benzofuran-3-one
23556
97%
97°C to 102°C
MFCD00051810
Light sensitive
Slightly miscible with water.
O=C1COC2=CC=CC=C12
134.13
134.13
3-Coumaranone
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only