missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroanisole, 98+%, Thermo Scientific Chemicals

57.29 € - 198.00 €

Chemical Identifiers

CAS 2845-89-8
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00000591
InChI Key YUKILTJWFRTXGB-UHFFFAOYSA-N
Synonym 3-chloroanisole, m-chloroanisole, benzene, 1-chloro-3-methoxy, anisole, m-chloro, 3-chloroanisol, meta-chloroanisole, 3-anisolyl chloride, pubchem3615, 3-chloromethoxybenzene, m-methoxyphenyl chloride
PubChem CID 17833
IUPAC Name 1-chloro-3-methoxybenzene
SMILES COC1=CC(=CC=C1)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11443567
View Documents
Thermo Scientific Alfa Aesar
A17327.18
50 g
57.29 €
50g
Please to purchase this item. Need a web account? Register with us today!
11453567
View Documents
Thermo Scientific Alfa Aesar
A17327.30
250 g
198.00 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2845-89-8
142.582
YUKILTJWFRTXGB-UHFFFAOYSA-N
17833
COC1=CC(=CC=C1)Cl
C7H7ClO
MFCD00000591
3-chloroanisole, m-chloroanisole, benzene, 1-chloro-3-methoxy, anisole, m-chloro, 3-chloroanisol, meta-chloroanisole, 3-anisolyl chloride, pubchem3615, 3-chloromethoxybenzene, m-methoxyphenyl chloride
1-chloro-3-methoxybenzene

Specifications

2845-89-8
192°C to 194°C
C7H7ClO
MFCD00000591
3-chloroanisole, m-chloroanisole, benzene, 1-chloro-3-methoxy, anisole, m-chloro, 3-chloroanisol, meta-chloroanisole, 3-anisolyl chloride, pubchem3615, 3-chloromethoxybenzene, m-methoxyphenyl chloride
COC1=CC(=CC=C1)Cl
142.582
142.59
3-Chloroanisole
1.168
73°C (163°F)
1.536
2041497
YUKILTJWFRTXGB-UHFFFAOYSA-N
1-chloro-3-methoxybenzene
17833
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

missing translation for 'einecsNumber' : 220-642-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.