Learn More
3-Chloro-N-methylaniline, 97%, Thermo Scientific Chemicals
24.80 € - 329.00 €
Chemical Identifiers
CAS | 7006-52-2 |
---|---|
Molecular Formula | C7H8ClN |
Molecular Weight (g/mol) | 141.60 |
MDL Number | MFCD00052014 |
InChI Key | WFGYSQDPURFIFL-UHFFFAOYSA-N |
Synonym | n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline |
PubChem CID | 138900 |
IUPAC Name | 3-chloro-N-methylaniline |
SMILES | CNC1=CC=CC(Cl)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11479603
|
Thermo Scientific Alfa Aesar
A12418.03 |
1 g | |||||||
11489603
|
Thermo Scientific Alfa Aesar
A12418.06 |
5 g |
87.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499603
|
Thermo Scientific Alfa Aesar
A12418.14 |
25 g |
329.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7006-52-2 | |
141.60 | |
WFGYSQDPURFIFL-UHFFFAOYSA-N | |
138900 | |
CNC1=CC=CC(Cl)=C1 |
C7H8ClN | |
MFCD00052014 | |
n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline | |
3-chloro-N-methylaniline |
Specifications
7006-52-2 | |
243°C to 244°C | |
C7H8ClN | |
MFCD00052014 | |
2802319 | |
WFGYSQDPURFIFL-UHFFFAOYSA-N | |
3-chloro-N-methylaniline | |
138900 | |
97% |
1.163 | |
>110°C (230°F) | |
1.583 | |
UN2810 | |
n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline | |
CNC1=CC=CC(Cl)=C1 | |
141.60 | |
141.6 | |
3-Chloro-N-methylaniline |
Safety and Handling
GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only