missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5,5-dimethyl-2-cyclohexen-1-one, 96%, Thermo Scientific Chemicals

Product Code. 11489592
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17530-69-7
Molecular Formula C8H11ClO
Molecular Weight (g/mol) 158.63
MDL Number MFCD00051620
InChI Key PJYTYJGMJDIKEJ-UHFFFAOYSA-N
Synonym 3-chloro-5,5-dimethyl-2-cyclohexen-1-one, 3-chloro-5,5-dimethylcyclohex-2-enone, unii-e73w928g5o, 3-chloro-5,5-dimethyl-2-cyclohexenone, 2-cyclohexen-1-one,3-chloro-5,5-dimethyl, 2-cyclohexen-1-one, 3-chloro-5,5-dimethyl, acmc-1bo45, 5,5-dimethyl-3-chlorocyclohex-2-enone, 3-chloro-5,5-dimethyl-cyclohex-2-enone, 3-chloro-5,5-dimethyl-1-cyclohex-2-enone
PubChem CID 140243
IUPAC Name 3-chloro-5,5-dimethylcyclohex-2-en-1-one
SMILES CC1(C)CC(Cl)=CC(=O)C1

Specifications

Density 1.084
Boiling Point 44°C to 46°C (0.1 mmHg)
Quantity 5 g
Beilstein 1099253
Formula Weight 158.63
Percent Purity 96%
Chemical Name or Material 3-Chloro-5,5-dimethyl-2-cyclohexen-1-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'