missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4'-fluoropropiophenone, 97%, Thermo Scientific Chemicals

27.30 € - 222.00 €

Chemical Identifiers

CAS 347-93-3
Molecular Formula C9H8ClFO
Molecular Weight (g/mol) 186.61
MDL Number MFCD00000991
InChI Key AAHQPLJUSLMHHR-UHFFFAOYSA-N
Synonym 3-chloro-4'-fluoropropiophenone, 3-chloro-1-4-fluorophenyl propan-1-one, 3-chloro-1-4-fluorophenyl-1-propanone, 1-propanone, 3-chloro-1-4-fluorophenyl, p-fluoro-, a-chloropropiophenone, pubchem13250, acmc-1ctxg, ksc573a4j, 3-chloro-4'fluoropropiophenone, 3-chloro4'-fluoropropiophenone
PubChem CID 67672
IUPAC Name 3-chloro-1-(4-fluorophenyl)propan-1-one
SMILES C1=CC(=CC=C1C(=O)CCCl)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11484957
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B21891.06
5 g
27.30 €
5g
Please to purchase this item. Need a web account? Register with us today!
11494957
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B21891.14
25 g
77.40 €
25g
Please to purchase this item. Need a web account? Register with us today!
11359499
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B21891.22
100 g
222.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

347-93-3
186.61
AAHQPLJUSLMHHR-UHFFFAOYSA-N
67672
C1=CC(=CC=C1C(=O)CCCl)F
C9H8ClFO
MFCD00000991
3-chloro-4'-fluoropropiophenone, 3-chloro-1-4-fluorophenyl propan-1-one, 3-chloro-1-4-fluorophenyl-1-propanone, 1-propanone, 3-chloro-1-4-fluorophenyl, p-fluoro-, a-chloropropiophenone, pubchem13250, acmc-1ctxg, ksc573a4j, 3-chloro-4'fluoropropiophenone, 3-chloro4'-fluoropropiophenone
3-chloro-1-(4-fluorophenyl)propan-1-one

Specifications

347-93-3
100°C to 103°C (3 mmHg)
MFCD00000991
3-chloro-4'-fluoropropiophenone, 3-chloro-1-4-fluorophenyl propan-1-one, 3-chloro-1-4-fluorophenyl-1-propanone, 1-propanone, 3-chloro-1-4-fluorophenyl, p-fluoro-, a-chloropropiophenone, pubchem13250, acmc-1ctxg, ksc573a4j, 3-chloro-4'fluoropropiophenone, 3-chloro4'-fluoropropiophenone
C1=CC(=CC=C1C(=O)CCCl)F
186.61
186.61
3-Chloro-4'-fluoropropiophenone
46°C to 49°C
C9H8ClFO
1866785
AAHQPLJUSLMHHR-UHFFFAOYSA-N
3-chloro-1-(4-fluorophenyl)propan-1-one
67672
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 206-475-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only