missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-cyanopyridine, 95%, Thermo Scientific™

118.00 € - 294.00 €

Chemical Identifiers

CAS 38180-46-0
Molecular Formula C6H3ClN2
Molecular Weight (g/mol) 138.55
MDL Number MFCD03788834
InChI Key YDPLFBIGFQFIDB-UHFFFAOYSA-N
Synonym 3-chloro-2-cyanopyridine, 2-cyano-3-chloropyridine, 3-chloropicolinonitrile, 3-chloro-2-pyridinecarbonitrile, 2-pyridinecarbonitrile, 3-chloro, 3-chloro-pyridine-2-carbonitrile, 3-chlorpyridin-2-carbonitril, chloro-2-cyanopyridine, pubchem14368, acmc-209iyb
PubChem CID 818258
IUPAC Name 3-chloropyridine-2-carbonitrile
SMILES ClC1=C(N=CC=C1)C#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15423567
SDS View Documents
Thermo Scientific Alfa Aesar
H26932.MD
250 mg N/A N/A N/A
11421520
SDS View Documents
Thermo Scientific Alfa Aesar
H26932.03
1 g N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

38180-46-0
138.55
YDPLFBIGFQFIDB-UHFFFAOYSA-N
818258
ClC1=C(N=CC=C1)C#N
C6H3ClN2
MFCD03788834
3-chloro-2-cyanopyridine, 2-cyano-3-chloropyridine, 3-chloropicolinonitrile, 3-chloro-2-pyridinecarbonitrile, 2-pyridinecarbonitrile, 3-chloro, 3-chloro-pyridine-2-carbonitrile, 3-chlorpyridin-2-carbonitril, chloro-2-cyanopyridine, pubchem14368, acmc-209iyb
3-chloropyridine-2-carbonitrile

Specifications

38180-46-0
C6H3ClN2
UN3439
YDPLFBIGFQFIDB-UHFFFAOYSA-N
3-chloropyridine-2-carbonitrile
818258
95%
83°C to 85°C
MFCD03788834
3-chloro-2-cyanopyridine, 2-cyano-3-chloropyridine, 3-chloropicolinonitrile, 3-chloro-2-pyridinecarbonitrile, 2-pyridinecarbonitrile, 3-chloro, 3-chloro-pyridine-2-carbonitrile, 3-chlorpyridin-2-carbonitril, chloro-2-cyanopyridine, pubchem14368, acmc-209iyb
ClC1=C(N=CC=C1)C#N
138.55
138.56
3-Chloro-2-cyanopyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c

H302+H332-H311-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.