missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Bromomethyl)-5-chlorobenzo[b]thiophene, 95+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 261.57 |
Percent Purity | 95% |
Chemical Name or Material | 3-(Bromomethyl)-5-chlorobenzo[b]thiophene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction