Learn More
3-Bromobenzo[b]thiophene, 95%, Thermo Scientific Chemicals
82.80 € - 254.00 €
Chemical Identifiers
CAS | 7342-82-7 |
---|---|
Molecular Formula | C8H5BrS |
Molecular Weight (g/mol) | 213.092 |
MDL Number | MFCD00023009 |
InChI Key | SRWDQSRTOOMPMO-UHFFFAOYSA-N |
Synonym | 3-bromobenzo b thiophene, 3-bromothianaphthene, 3-bromobenzothiophene, benzo b thiophene, 3-bromo, 3-bromo-benzo b thiophene, 3-bromo benzothiophene, benzo b thiophene, 3-bromo-,, zlchem 804 |
PubChem CID | 123250 |
IUPAC Name | 3-bromo-1-benzothiophene |
SMILES | C1=CC=C2C(=C1)C(=CS2)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15425737
|
Thermo Scientific Alfa Aesar
H32948.09 |
10 g |
82.80 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15435737
|
Thermo Scientific Alfa Aesar
H32948.18 |
50 g |
254.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Antifungal activity of synthetic di (hetero) arylamines based on the benzo [b] thiophene moiety.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAntifungal activity of synthetic di (hetero) arylamines based on the benzo [b] thiophene moiety.
Solubility
Sparingly Soluble (0.14 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
7342-82-7 | |
213.092 | |
SRWDQSRTOOMPMO-UHFFFAOYSA-N | |
123250 | |
C1=CC=C2C(=C1)C(=CS2)Br |
C8H5BrS | |
MFCD00023009 | |
3-bromobenzo b thiophene, 3-bromothianaphthene, 3-bromobenzothiophene, benzo b thiophene, 3-bromo, 3-bromo-benzo b thiophene, 3-bromo benzothiophene, benzo b thiophene, 3-bromo-,, zlchem 804 | |
3-bromo-1-benzothiophene |
Specifications
7342-82-7 | |
72°C to 74°C (0.27 mmHg) | |
1.668 | |
3-bromobenzo b thiophene, 3-bromothianaphthene, 3-bromobenzothiophene, benzo b thiophene, 3-bromo, 3-bromo-benzo b thiophene, 3-bromo benzothiophene, benzo b thiophene, 3-bromo-,, zlchem 804 | |
SRWDQSRTOOMPMO-UHFFFAOYSA-N | |
3-bromo-1-benzothiophene | |
123250 | |
95% |
1.629 | |
C8H5BrS | |
MFCD00023009 | |
Sparingly Soluble (0.14g/L) (25°C). | |
C1=CC=C2C(=C1)C(=CS2)Br | |
213.092 | |
213.1 | |
3-Bromobenzo[b]thiophene |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
Recommended Storage : Ambient temperatures
RUO – Research Use Only