missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromoacetyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals

Product Code. 15575156
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17694-68-7
Molecular Formula C7H7Br2NO
Molecular Weight (g/mol) 280.95
MDL Number MFCD00052182
InChI Key WDTSYONULAZKIE-UHFFFAOYSA-N
Synonym 2-bromo-1-pyridin-3-yl ethanone hydrobromide, 3-bromoacetyl pyridine hydrobromide, 3-2-bromoacetyl pyridine hydrobromide, 2-bromo-1-3-pyridinyl ethanone hydrobromide, 3-bromoacetyl pyridinium bromide, 3-bromoacetyl pyridine hbr, 2-bromo-1-pyridin-3-ylethan-1-one hydrobromide, 2-bromo-1-pyridin-3-ylethanone hydrobromide, 3-bromoacetylpyridine hydrobromide
PubChem CID 2776234
IUPAC Name 2-bromo-1-pyridin-3-ylethanone;hydrobromide
SMILES [H+].[Br-].BrCC(=O)C1=CC=CN=C1

Specifications

Melting Point 192.0°C to 196.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Quantity 25 g
Formula Weight 280.94
Percent Purity 98%
Chemical Name or Material 3-(Bromoacetyl)pyridine hydrobromide
compliance-icons
Product Identifier
  • 3-(Bromoacetyl)pyridine, hydrobromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.