missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Bromo-5'-fluoroacetophenone, 96%, Thermo Scientific Chemicals
71.50 € - 855.00 €
Chemical Identifiers
CAS | 105515-20-6 |
---|---|
Molecular Formula | C8H6BrFO |
Molecular Weight (g/mol) | 217.04 |
MDL Number | MFCD11847746 |
InChI Key | MZDXPUDHZBCYGS-UHFFFAOYSA-N |
Synonym | 1-3-bromo-5-fluorophenyl ethanone, 3'-bromo-5'-fluoroacetophenone, 1-3-bromo-5-fluorophenyl ethan-1-one, ethanone, 1-3-bromo-5-fluorophenyl, 3-bromo-5-fluoroacetophenone, pubchem22227, 1-3-bromo-5-fluoro-phenyl-ethanone |
PubChem CID | 13730359 |
IUPAC Name | 1-(3-bromo-5-fluorophenyl)ethanone |
SMILES | CC(=O)C1=CC(F)=CC(Br)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15489408
|
Thermo Scientific Alfa Aesar
H62977.MD |
250 mg | |||||||
15469408
|
Thermo Scientific Alfa Aesar
H62977.03 |
1 g |
220.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15479408
|
Thermo Scientific Alfa Aesar
H62977.06 |
5 g |
855.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
105515-20-6 | |
217.04 | |
MZDXPUDHZBCYGS-UHFFFAOYSA-N | |
13730359 | |
CC(=O)C1=CC(F)=CC(Br)=C1 |
C8H6BrFO | |
MFCD11847746 | |
1-3-bromo-5-fluorophenyl ethanone, 3'-bromo-5'-fluoroacetophenone, 1-3-bromo-5-fluorophenyl ethan-1-one, ethanone, 1-3-bromo-5-fluorophenyl, 3-bromo-5-fluoroacetophenone, pubchem22227, 1-3-bromo-5-fluoro-phenyl-ethanone | |
1-(3-bromo-5-fluorophenyl)ethanone |
Specifications
105515-20-6 | |
MFCD11847746 | |
MZDXPUDHZBCYGS-UHFFFAOYSA-N | |
1-(3-bromo-5-fluorophenyl)ethanone | |
13730359 | |
96% | |
3'-Bromo-5'-fluoroacetophenone |
C8H6BrFO | |
1-3-bromo-5-fluorophenyl ethanone, 3'-bromo-5'-fluoroacetophenone, 1-3-bromo-5-fluorophenyl ethan-1-one, ethanone, 1-3-bromo-5-fluorophenyl, 3-bromo-5-fluoroacetophenone, pubchem22227, 1-3-bromo-5-fluoro-phenyl-ethanone | |
CC(=O)C1=CC(F)=CC(Br)=C1 | |
217.04 | |
217.04 | |
Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only