missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylthiophene, 95%, Thermo Scientific Chemicals

Product Code. 10217842
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30318-99-1
Molecular Formula C5H5BrS
Molecular Weight (g/mol) 177.06
MDL Number MFCD00130084
InChI Key MBUSOPVRLCFJCS-UHFFFAOYSA-N
Synonym 3-bromo-4-methyl-thiophene, thiophene, 3-bromo-4-methyl, pubchem21832, acmc-209her, 3-bromo-4-methylthiophen, 4-bromo-3-methylthiophene, bidd:gt0486, 3-bromo-4-methylthiophene
PubChem CID 2734935
IUPAC Name 3-bromo-4-methylthiophene
SMILES CC1=CSC=C1Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5840g/mL
Boiling Point 179°C to 183°C (770.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5770 to 1.579
Quantity 5 g
Specific Gravity 1.584
Formula Weight 177.06
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-methylthiophene
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-4-methylthiophene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.