missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-fluorocinnamic acid, 98%, Thermo Scientific™
44.60 € - 499.00 €
Chemical Identifiers
CAS | 160434-49-1 |
---|---|
Molecular Formula | C9H5BrFO2 |
Molecular Weight (g/mol) | 244.04 |
MDL Number | MFCD00143095 |
InChI Key | ZNIGVADAKXOMQH-DUXPYHPUSA-M |
Synonym | 3-bromo-4-fluorocinnamic acid, 2e-3-3-bromo-4-fluorophenyl prop-2-enoic acid, e-3-3-bromo-4-fluorophenyl acrylic acid, 3-3-bromo-4-fluoro-phenyl-acrylic acid, 3-3-bromo-4-fluorophenyl acrylic acid, pubchem3228, 2e-3-3-bromo-4-fluorophenyl acrylic acid, rarechem bk hw 0193, timtec-bb sbb003187, attercop-chm at116391 |
PubChem CID | 688312 |
IUPAC Name | (E)-3-(3-bromo-4-fluorophenyl)prop-2-enoic acid |
SMILES | [O-]C(=O)\C=C\C1=CC=C(F)C(Br)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11491878
|
Thermo Scientific Alfa Aesar
A14526.06 |
5 g |
44.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11401888
|
Thermo Scientific Alfa Aesar
A14526.14 |
25 g | |||||||
11411888
|
Thermo Scientific Alfa Aesar
A14526.22 |
100 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
160434-49-1 | |
244.04 | |
ZNIGVADAKXOMQH-DUXPYHPUSA-M | |
688312 | |
[O-]C(=O)\C=C\C1=CC=C(F)C(Br)=C1 |
C9H5BrFO2 | |
MFCD00143095 | |
3-bromo-4-fluorocinnamic acid, 2e-3-3-bromo-4-fluorophenyl prop-2-enoic acid, e-3-3-bromo-4-fluorophenyl acrylic acid, 3-3-bromo-4-fluoro-phenyl-acrylic acid, 3-3-bromo-4-fluorophenyl acrylic acid, pubchem3228, 2e-3-3-bromo-4-fluorophenyl acrylic acid, rarechem bk hw 0193, timtec-bb sbb003187, attercop-chm at116391 | |
(E)-3-(3-bromo-4-fluorophenyl)prop-2-enoic acid |
Specifications
160434-49-1 | |
C9H5BrFO2 | |
8977970 | |
ZNIGVADAKXOMQH-DUXPYHPUSA-M | |
(E)-3-(3-bromo-4-fluorophenyl)prop-2-enoic acid | |
688312 | |
98% |
193°C to 195°C | |
MFCD00143095 | |
3-bromo-4-fluorocinnamic acid, 2e-3-3-bromo-4-fluorophenyl prop-2-enoic acid, e-3-3-bromo-4-fluorophenyl acrylic acid, 3-3-bromo-4-fluoro-phenyl-acrylic acid, 3-3-bromo-4-fluorophenyl acrylic acid, pubchem3228, 2e-3-3-bromo-4-fluorophenyl acrylic acid, rarechem bk hw 0193, timtec-bb sbb003187, attercop-chm at116391 | |
[O-]C(=O)\C=C\C1=CC=C(F)C(Br)=C1 | |
244.04 | |
245.05 | |
3-Bromo-4-fluorocinnamic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures