missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-furoic acid, 97%, Thermo Scientific™

82.10 € - 230.00 €

Chemical Identifiers

CAS 14903-90-3
Molecular Formula C5H3BrO3
Molecular Weight (g/mol) 190.98
MDL Number MFCD00092761
InChI Key UZBGSJZFBUOJNE-UHFFFAOYSA-N
PubChem CID 605479
IUPAC Name 3-bromofuran-2-carboxylic acid
SMILES C1=COC(=C1Br)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10566704
View Documents
Thermo Scientific Maybridge
CC67201CB
250 mg Amber Glass Bottle
82.10 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10441884
View Documents
Thermo Scientific Maybridge
CC67201DA
1 g Amber Glass Bottle
230.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

14903-90-3
190.98
UZBGSJZFBUOJNE-UHFFFAOYSA-N
3-bromofuran-2-carboxylic acid
C5H3BrO3
MFCD00092761
605479
C1=COC(=C1Br)C(=O)O

Specifications

14903-90-3
100.0
C5H3BrO3
3-bromo-2-furoic acid, furancarboxylic acid, bromo, 3-bromo-2-furancarboxylic acid, 3-bromo-furan-2-carboxylic acid, 3-bromofuran-2-carboxylicacid, bromofuroic acid, 3-bomo-2-furoic acid, 2-furancarboxylic acid,3-bromo, 3-bromanylfuran-2-carboxylic acid, 2-furancarboxylic acid, 3-bromo
C1=COC(=C1Br)C(=O)O
190.98
190.98
3-Bromo-2-furoic acid
96.5
97%
MFCD00092761
UZBGSJZFBUOJNE-UHFFFAOYSA-N
3-bromofuran-2-carboxylic acid
605479
97%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.