missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-cyanobenzeneboronic acid, 96%, Thermo Scientific Chemicals

Product Code. 15408468
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1032231-32-5
Molecular Formula C7H5BBrNO2
Molecular Weight (g/mol) 225.836
InChI Key LHGIIQFOQAPPQA-UHFFFAOYSA-N
Synonym 3-bromo-2-cyanophenyl boronic acid, 2-cyano-3-bromophenylboronic acid
PubChem CID 46739320
IUPAC Name (3-bromo-2-cyanophenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)Br)C#N)(O)O

Specifications

Quantity 1 g
Formula Weight 225.84
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Bromo-2-cyanobenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.