missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenylboronic acid monohydrate, 98%, Thermo Scientific Chemicals

51.10 € - 163.00 €

Chemical Identifiers

CAS 206658-89-1
Molecular Formula C6H10BNO3
Molecular Weight (g/mol) 154.96
MDL Number MFCD00149554
InChI Key XAEOVQODHLLNKX-UHFFFAOYSA-N
Synonym 3-aminophenylboronic acid monohydrate, 3-aminophenyl boronic acid hydrate, 3-aminophenylboronic acid hydrate, 3-aminobenzeneboronic acid monohydrate, m-aminophenylboronic acid hydrate, boronic acid, 3-aminophenyl-, monohydrate, 3-aminobenzeneboronic acid hydrate, pubchem4016, acmc-1cfgg, 3-boronoaniline monohydrate
PubChem CID 14389423
IUPAC Name (3-aminophenyl)boronic acid;hydrate
SMILES O.NC1=CC=CC(=C1)B(O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10726062
View Documents Promotion Details
Thermo Scientific Acros
352760010
1 g Glass bottle
51.10 €
1g
Please to purchase this item. Need a web account? Register with us today!
10579643
View Documents Promotion Details
Thermo Scientific Acros
352760050
5 g Glass bottle
163.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

206658-89-1
154.96
XAEOVQODHLLNKX-UHFFFAOYSA-N
14389423
O.NC1=CC=CC(=C1)B(O)O
C6H10BNO3
MFCD00149554
3-aminophenylboronic acid monohydrate, 3-aminophenyl boronic acid hydrate, 3-aminophenylboronic acid hydrate, 3-aminobenzeneboronic acid monohydrate, m-aminophenylboronic acid hydrate, boronic acid, 3-aminophenyl-, monohydrate, 3-aminobenzeneboronic acid hydrate, pubchem4016, acmc-1cfgg, 3-boronoaniline monohydrate
(3-aminophenyl)boronic acid;hydrate

Specifications

206658-89-1
100.0
Beige to Brown
98%
NH2C6H4B(OH)2·H2O
3-aminophenylboronic acid monohydrate, 3-aminophenyl boronic acid hydrate, 3-aminophenylboronic acid hydrate, 3-aminobenzeneboronic acid monohydrate, m-aminophenylboronic acid hydrate, boronic acid, 3-aminophenyl-, monohydrate, 3-aminobenzeneboronic acid hydrate, pubchem4016, acmc-1cfgg, 3-boronoaniline monohydrate
XAEOVQODHLLNKX-UHFFFAOYSA-N
(3-aminophenyl)boronic acid;hydrate
14389423
98%
3-Aminophenylboronic acid monohydrate
97.5
94°C to 97°C
Authentic
C6H10BNO3
MFCD00149554
Solubility in water: soluble
O.NC1=CC=CC(=C1)B(O)O
154.96
154.97
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

Product Identifier
  • 3-Aminophenylboronic acid monohydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only