missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzonitrile, 99%, Thermo Scientific Chemicals

26.13 € - 279.39 €

Chemical Identifiers

CAS 2237-30-1
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00007756
InChI Key NJXPYZHXZZCTNI-UHFFFAOYSA-N
Synonym 3-cyanoaniline, m-aminobenzonitrile, m-cyanoaniline, 3-amino-benzonitrile, 3-aminobenzonitril, benzonitrile, m-amino, m-anthranilonitrile, benzonitrile, 3-amino, unii-15v3cw939x, 3-amino benzonitrile
PubChem CID 16702
IUPAC Name 3-aminobenzonitrile
SMILES C1=CC(=CC(=C1)N)C#N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10381604
SDS View Documents
Thermo Scientific Acros
173040050
5 g Glass bottle
26.13 €
5g
Please to purchase this item. Need a web account? Register with us today!
10694662
SDS View Documents
Thermo Scientific Acros
173040250
25 g Glass bottle
81.10 €
25g
Please to purchase this item. Need a web account? Register with us today!
10391604
SDS View Documents
Thermo Scientific Acros
173041000
100 g Glass bottle
279.39 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2237-30-1
118.14
NJXPYZHXZZCTNI-UHFFFAOYSA-N
16702
C1=CC(=CC(=C1)N)C#N
C7H6N2
MFCD00007756
3-cyanoaniline, m-aminobenzonitrile, m-cyanoaniline, 3-amino-benzonitrile, 3-aminobenzonitril, benzonitrile, m-amino, m-anthranilonitrile, benzonitrile, 3-amino, unii-15v3cw939x, 3-amino benzonitrile
3-aminobenzonitrile

Specifications

2237-30-1
100.0
Brown
112°C
99%
H2NC6H4CN
14, 391
Solubility in water: insoluble. Other solubilities: soluble in ehtanol,ethyl ether,acetone
C1=CC(=CC(=C1)N)C#N
118.14
118.14
Crystalline Solid
98.5
50.0°C to 54.0°C
288.0°C to 290.0°C
Authentic
C7H6N2
MFCD00007756
3-cyanoaniline, m-aminobenzonitrile, m-cyanoaniline, 3-amino-benzonitrile, 3-aminobenzonitril, benzonitrile, m-amino, m-anthranilonitrile, benzonitrile, 3-amino, unii-15v3cw939x, 3-amino benzonitrile
NJXPYZHXZZCTNI-UHFFFAOYSA-N
3-aminobenzonitrile
16702
99%
3-Aminobenzonitrile, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POI

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 218-800-5

missing translation for 'rtecsNumber' : DI2454000

missing translation for 'tsca' : TSCA

Product Identifier
  • 3-Aminobenzonitrile

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Aminobenzonitrile, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.