missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-chlorobenzeneboronic acid hydrochloride, 95%, Thermo Scientific™

Product Code. 15400186
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850568-45-5
Molecular Formula C6H8BCl2NO2
Molecular Weight (g/mol) 207.845
MDL Number MFCD04115643
InChI Key MTMLYYSBEQYCDJ-UHFFFAOYSA-N
Synonym 3-amino-4-chlorophenyl boronic acid hydrochloride, 3-amino-4-chlorophenylboronic acid, hcl, 3-amino-4-chlorophenylboronic acid hydrochloride, 3-amino-4-chlorophenyl boronicacidhydrochloride, 3-amino-4-chlorobenzeneboronic acid hydrochloride, 3-amino-4-chlorophenylboronic acid hcl, 3-amino-4-chlorophenyl boronic acid-hydrogen chloride 1/1
PubChem CID 44118731
IUPAC Name (3-amino-4-chlorophenyl)boronic acid;hydrochloride
SMILES B(C1=CC(=C(C=C1)Cl)N)(O)O.Cl

Specifications

Melting Point 278°C to 288°C
Quantity 1 g
Formula Weight 207.85
Percent Purity 95%
Chemical Name or Material 3-Amino-4-chlorobenzeneboronic acid hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.