missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-bromo-5-fluoropyridine, 97%, Thermo Scientific Chemicals

Product Code. 15446677
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 884495-03-8
Molecular Formula C5H4BrFN2
Molecular Weight (g/mol) 191.00
MDL Number MFCD05662413
InChI Key QUZAKZBKMMUARE-UHFFFAOYSA-N
Synonym 3-amino-2-bromo-5-fluoropyridine, 2-bromo-5-fluoro-3-pyridylamine, 2-bromo-3-amino-5-fluoropyridine, 2-bromo-5-fluoro-3-aminopyridine, 3-pyridinamine, 2-bromo-5-fluoro, abbypharma ap-31-3805, 2-bromo-5-fluoro-3-pyridinamine, 2-bromo-5-fluoro-pyridin-3-ylamine, pubchem1145, acmc-209qtb
PubChem CID 40427367
IUPAC Name 2-bromo-5-fluoropyridin-3-amine
SMILES NC1=CC(F)=CN=C1Br

Specifications

Melting Point 97°C to 101°C
Quantity 1 g
Formula Weight 191
Percent Purity 97%
Chemical Name or Material 3-Amino-2-bromo-5-fluoropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.