missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylpyridine, 98%, Thermo Scientific Chemicals

28.44 € - 414.06 €

Chemical Identifiers

CAS 350-03-8
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
MDL Number MFCD00006396
InChI Key WEGYGNROSJDEIW-UHFFFAOYSA-N
Synonym 3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one
PubChem CID 9589
IUPAC Name 1-pyridin-3-ylethanone
SMILES CC(=O)C1=CN=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10080600
View Documents
Thermo Scientific Acros
102690100
10 g Glass bottle
28.44 €
10g
Please to purchase this item. Need a web account? Register with us today!
10090600
View Documents
Thermo Scientific Acros
102691000
100 g Glass bottle
124.62 €
100g
Please to purchase this item. Need a web account? Register with us today!
10592534
View Documents
Thermo Scientific Acros
102695000
500 g Glass bottle
414.06 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

350-03-8
121.14
WEGYGNROSJDEIW-UHFFFAOYSA-N
9589
CC(=O)C1=CN=CC=C1
C7H7NO
MFCD00006396
3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one
1-pyridin-3-ylethanone

Specifications

350-03-8
100.0
Colorless to Yellow
220.0°C
Authentic
C7H7NO
1.5320 to 1.5350
21,279
1.1
Solubility in water: soluble in hot water. Other solubilities: soluble in acids,miscible with organic solvents except ligroin
CC(=O)C1=CN=CC=C1
121.14
121.14
Liquid
97.5
12.0°C to 13.0°C
1.1000g/mL
104°C
97.5% min. (GC)
(C5H4N)COCH3
MFCD00006396
15,6189
3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one
WEGYGNROSJDEIW-UHFFFAOYSA-N
1-pyridin-3-ylethanone
9589
98%
3-Acetylpyridine, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Fatal if swallowed.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiou

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 206-496-7

Product Identifier
  • 3-Acetylpyridine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.