Learn More
3-Acetylpyridine, 98%, Thermo Scientific Chemicals
42.50 € - 466.00 €
Chemical Identifiers
CAS | 350-03-8 |
---|---|
Molecular Formula | C7H7NO |
Molecular Weight (g/mol) | 121.139 |
MDL Number | MFCD00006396 |
InChI Key | WEGYGNROSJDEIW-UHFFFAOYSA-N |
Synonym | 3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one |
PubChem CID | 9589 |
IUPAC Name | 1-pyridin-3-ylethanone |
SMILES | CC(=O)C1=CN=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11402377
|
Thermo Scientific Alfa Aesar
A14246.14 |
25 g |
42.50 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11412377
|
Thermo Scientific Alfa Aesar
A14246.22 |
100 g |
101.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11422377
|
Thermo Scientific Alfa Aesar
A14246.36 |
500 g |
466.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Acetylpyridine used as an intermediate for the synthesis of risedronate sodium, inhibitor of bone resorption. It is also used in perfumery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Acetylpyridine used as an intermediate for the synthesis of risedronate sodium, inhibitor of bone resorption. It is also used in perfumery.
Solubility
Soluble in Chloroform. Miscible with water.
Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
Chemical Identifiers
350-03-8 | |
121.139 | |
WEGYGNROSJDEIW-UHFFFAOYSA-N | |
9589 | |
CC(=O)C1=CN=CC=C1 |
C7H7NO | |
MFCD00006396 | |
3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one | |
1-pyridin-3-ylethanone |
Specifications
350-03-8 | |
1.106 | |
104°C (219°F) | |
C7H7NO | |
MFCD00006396 | |
107751 | |
3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one | |
WEGYGNROSJDEIW-UHFFFAOYSA-N | |
1-pyridin-3-ylethanone | |
9589 | |
98% |
12°C to 13°C | |
218°C to 220°C | |
Characteristic | |
1.535 | |
UN2810 | |
14,6116 | |
Soluble in Chloroform. Miscible with water. | |
CC(=O)C1=CN=CC=C1 | |
121.139 | |
121.14 | |
3-Acetylpyridine |
Safety and Handling
GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H300-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 206-496-7
missing translation for 'rtecsNumber' : OB5425000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.