missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-6-bromopyridine, 97%, Thermo Scientific Chemicals

Product Code. 15465997
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139042-59-4
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD04974527
InChI Key MUKKGHQBUKOMTD-UHFFFAOYSA-N
Synonym 5-acetyl-2-bromopyridine, 1-6-bromopyridin-3-yl ethanone, 2-bromo-5-acetylpyridine, 1-6-bromo-pyridin-3-yl-ethanone, 1-6-bromopyridin-3-yl ethan-1-one, 3-acetyl-6-bromopyridine, ethanone, 1-6-bromo-3-pyridinyl, 1-6-bromopyrid-3-yl ethanone, 1-6-bromo-3-pyridyl ethanone, acmc-209cij
PubChem CID 15668195
IUPAC Name 1-(6-bromopyridin-3-yl)ethanone
SMILES CC(=O)C1=CN=C(C=C1)Br

Specifications

Melting Point 124°C to 128°C
Quantity 1 g
Formula Weight 200.03
Percent Purity 97%
Chemical Name or Material 3-Acetyl-6-bromopyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'