missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetoxy-1-propenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
90.60 € - 245.00 €
Chemical Identifiers
CAS | 161395-97-7 |
---|---|
Molecular Formula | C11H19BO4 |
Molecular Weight (g/mol) | 226.08 |
MDL Number | MFCD03788751 |
InChI Key | ZXBRLGZFCXGTBA-UHFFFAOYSA-N |
Synonym | 3-acetoxy-1-propenylboronic acid pinacol ester, e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl allyl acetate, 2-3-acetoxy-1-propenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-propen-1-yl acetate, 2e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl prop-2-en-1-yl acetate, e-3-acetoxy-1-propenylboronic acid 2,3-dimethylbutane-2,3-diyl ester |
PubChem CID | 11264613 |
IUPAC Name | [(E)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)prop-2-enyl] acetate |
SMILES | CC(=O)OCC=CB1OC(C)(C)C(C)(C)O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11385535
|
Thermo Scientific Alfa Aesar
L19700.MD |
250 mg |
90.60 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11375535
|
Thermo Scientific Alfa Aesar
L19700.03 |
1 g |
245.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
161395-97-7 | |
226.08 | |
ZXBRLGZFCXGTBA-UHFFFAOYSA-N | |
11264613 | |
CC(=O)OCC=CB1OC(C)(C)C(C)(C)O1 |
C11H19BO4 | |
MFCD03788751 | |
3-acetoxy-1-propenylboronic acid pinacol ester, e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl allyl acetate, 2-3-acetoxy-1-propenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-propen-1-yl acetate, 2e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl prop-2-en-1-yl acetate, e-3-acetoxy-1-propenylboronic acid 2,3-dimethylbutane-2,3-diyl ester | |
[(E)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)prop-2-enyl] acetate |
Specifications
161395-97-7 | |
1.4485 | |
6923397 | |
ZXBRLGZFCXGTBA-UHFFFAOYSA-N | |
[(E)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)prop-2-enyl] acetate | |
11264613 | |
97% |
C11H19BO4 | |
MFCD03788751 | |
3-acetoxy-1-propenylboronic acid pinacol ester, e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl allyl acetate, 2-3-acetoxy-1-propenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-propen-1-yl acetate, 2e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl prop-2-en-1-yl acetate, e-3-acetoxy-1-propenylboronic acid 2,3-dimethylbutane-2,3-diyl ester | |
CC(=O)OCC=CB1OC(C)(C)C(C)(C)O1 | |
226.08 | |
226.08 | |
3-Acetoxy-1-propenylboronic acid pinacol ester |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only