missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

61.90 € - 234.00 €

Chemical Identifiers

CAS 192182-54-0
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.982
MDL Number MFCD03095127
InChI Key XUIURRYWQBBCCK-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzeneboronic acid, 3,5-dimethoxyphenyl boronic acid, 3,5-dimethoxyphenylboronicacid, boronic acid, 3,5-dimethoxyphenyl, pubchem1827, acmc-1c82c, ksc174i6h, 3,5-dimethoxy phenyl boronic acid, 3,5-dimethoxyphenylboronic acid, boronic acid, b-3,5-dimethoxyphenyl
PubChem CID 4374257
IUPAC Name (3,5-dimethoxyphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)OC)OC)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15409737
View Documents
Thermo Scientific Alfa Aesar
H52965.03
1 g
61.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
15419737
View Documents
Thermo Scientific Alfa Aesar
H52965.06
5 g
234.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

3,5-Dimethoxybenzeneboronic acid is used as reactant for palladium-catalyzed coupling reactions, preparation of benzopyranone derivatives as positive GABAA receptor modulators, preparation of aryl alkenes via three-component coupling catalyzed by palladium and Suzuki-Miyaura coupling. It is also used as reactant for rhodium catalyzed cyanation with N-cyano-N-phenyl-p-methylbenzenesulfonamide and preparation of bisphosphonate inhibitors of human farnesyl pyrophosphate synthase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethoxybenzeneboronic acid is used as reactant for palladium-catalyzed coupling reactions, preparation of benzopyranone derivatives as positive GABAA receptor modulators, preparation of aryl alkenes via three-component coupling catalyzed by palladium and Suzuki-Miyaura coupling. It is also used as reactant for rhodium catalyzed cyanation with N-cyano-N-phenyl-p-methylbenzenesulfonamide and preparation of bisphosphonate inhibitors of human farnesyl pyrophosphate synthase.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

192182-54-0
181.982
XUIURRYWQBBCCK-UHFFFAOYSA-N
4374257
B(C1=CC(=CC(=C1)OC)OC)(O)O
C8H11BO4
MFCD03095127
3,5-dimethoxybenzeneboronic acid, 3,5-dimethoxyphenyl boronic acid, 3,5-dimethoxyphenylboronicacid, boronic acid, 3,5-dimethoxyphenyl, pubchem1827, acmc-1c82c, ksc174i6h, 3,5-dimethoxy phenyl boronic acid, 3,5-dimethoxyphenylboronic acid, boronic acid, b-3,5-dimethoxyphenyl
(3,5-dimethoxyphenyl)boronic acid

Specifications

192182-54-0
C8H11BO4
3,5-dimethoxybenzeneboronic acid, 3,5-dimethoxyphenyl boronic acid, 3,5-dimethoxyphenylboronicacid, boronic acid, 3,5-dimethoxyphenyl, pubchem1827, acmc-1c82c, ksc174i6h, 3,5-dimethoxy phenyl boronic acid, 3,5-dimethoxyphenylboronic acid, boronic acid, b-3,5-dimethoxyphenyl
XUIURRYWQBBCCK-UHFFFAOYSA-N
(3,5-dimethoxyphenyl)boronic acid
4374257
98%
205°C to 207°C
MFCD03095127
Soluble in methanol.
B(C1=CC(=CC(=C1)OC)OC)(O)O
181.982
181.98
3,5-Dimethoxybenzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.