missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethoxy-4-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
3,5-Dimethoxy-4-methylbenzoic acid undergoes reduction in the presence of sodium in ethanol to prepare 1,4-dihydro-3,5-dimethoxy- kmethylbenzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dimethoxy-4-methylbenzoic acid undergoes reduction in the presence of sodium in ethanol to prepare 1,4-dihydro-3,5-dimethoxy- kmethylbenzoic acid.
Solubility
Soluble in liquid ammonia.
Notes
Incompatible with oxidizing agents.
3,5-Dimethoxy-4-methylbenzoic acid undergoes reduction in the presence of sodium in ethanol to prepare 1,4-dihydro-3,5-dimethoxy- kmethylbenzoic acid.
Solubility
Soluble in liquid ammonia.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 210°C to 216°C |
Quantity | 25 g |
Beilstein | 2695238 |
Solubility Information | Soluble in liquid ammonia. |
Formula Weight | 196.2 |
Percent Purity | 97% |
Chemical Name or Material | 3,5-Dimethoxy-4-methylbenzoic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction