missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenylacetic acid, 95%, Thermo Scientific Chemicals

Product Code. 15409928
Click to view available options
Quantity:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 51719-65-4
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD01764671
InChI Key RERINLRFXYGZEE-UHFFFAOYSA-N
Synonym 2-3,5-dichlorophenyl acetic acid, 3,5-dichlorophenylacetic acid, benzeneacetic acid, 3,5-dichloro, 3,5-dichlorobenzeneacetic acid, 3,5-dichlorophenyl acetic acid, 3,5-dichloro-phenyl-acetic acid, 2-3,5-dichlorophenyl-acetic acid, 3,5-dichlorophenylaceticacid, pubchem24116, 3,5-dichlorphenylacetic acid
PubChem CID 6452487
IUPAC Name 2-(3,5-dichlorophenyl)acetic acid
SMILES C1=C(C=C(C=C1Cl)Cl)CC(=O)O

Spezifikation

Quantity 250 mg
Formula Weight 205.03
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3,5-Dichlorophenylacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'