missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloroaniline, 98%, Thermo Scientific Chemicals

48.14 € - 148.74 €

Chemical Identifiers

CAS 626-43-7
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.02
MDL Number MFCD00007774
InChI Key UQRLKWGPEVNVHT-UHFFFAOYSA-N
Synonym benzenamine, 3,5-dichloro, m-dichloroaniline, aniline, 3,5-dichloro, 3,5-dichlorobenzenamine, 3,5-dichloro benzenamine, unii-oz75zm1s3g, 3,5-dichloranilin, 3,5 dichloraniline, ccris 2396, 3,5-dichloro aniline
PubChem CID 12281
ChEBI CHEBI:19904
IUPAC Name 3,5-dichloroaniline
SMILES C1=C(C=C(C=C1Cl)Cl)N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10429600
View Documents
Thermo Scientific Acros
113121000
100 g Glass bottle
48.14 €
100g
Please to purchase this item. Need a web account? Register with us today!
10030810
View Documents
Thermo Scientific Acros
113125000
500 g Glass bottle
148.74 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

626-43-7
162.02
UQRLKWGPEVNVHT-UHFFFAOYSA-N
12281
3,5-dichloroaniline
C6H5Cl2N
MFCD00007774
benzenamine, 3,5-dichloro, m-dichloroaniline, aniline, 3,5-dichloro, 3,5-dichlorobenzenamine, 3,5-dichloro benzenamine, unii-oz75zm1s3g, 3,5-dichloranilin, 3,5 dichloraniline, ccris 2396, 3,5-dichloro aniline
CHEBI:19904
C1=C(C=C(C=C1Cl)Cl)N

Specifications

626-43-7
100.0
White to Brown
260.0°C (741.0 mmHg)
Authentic
C6H5Cl2N
MFCD00007774
1.58
Solubility in water: 0.6g/L (26°C). Other solubilities: 26.8g/L n-hexane (20°C),soluble in most organic solvents
C1=C(C=C(C=C1Cl)Cl)N
162.02
CHEBI:19904
162.02
Crystals
97.5
49.0°C to 53.0°C
1.5800g/mL
133°C
97.5% min. (GC)
Cl2C6H3NH2
12, 626
benzenamine, 3,5-dichloro, m-dichloroaniline, aniline, 3,5-dichloro, 3,5-dichlorobenzenamine, 3,5-dichloro benzenamine, unii-oz75zm1s3g, 3,5-dichloranilin, 3,5 dichloraniline, ccris 2396, 3,5-dichloro aniline
UQRLKWGPEVNVHT-UHFFFAOYSA-N
3,5-dichloroaniline
12281
2.1 mPa.s (50°C)
98%
3, 5-Dichloroaniline, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 210-948-9

missing translation for 'tsca' : TSCA

Recommended Storage : Product darkens on storage

Product Identifier
  • 3,5-Dichloroaniline

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.