Learn More
3,5-Dichloroaniline, 98%, Thermo Scientific Chemicals
30.00 € - 357.00 €
Chemical Identifiers
CAS | 626-43-7 |
---|---|
Molecular Formula | C6H5Cl2N |
Molecular Weight (g/mol) | 162.013 |
MDL Number | MFCD00007774 |
InChI Key | UQRLKWGPEVNVHT-UHFFFAOYSA-N |
Synonym | benzenamine, 3,5-dichloro, m-dichloroaniline, aniline, 3,5-dichloro, 3,5-dichlorobenzenamine, 3,5-dichloro benzenamine, unii-oz75zm1s3g, 3,5-dichloranilin, 3,5 dichloraniline, ccris 2396, 3,5-dichloro aniline |
PubChem CID | 12281 |
ChEBI | CHEBI:19904 |
IUPAC Name | 3,5-dichloroaniline |
SMILES | C1=C(C=C(C=C1Cl)Cl)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11491638
|
Thermo Scientific Alfa Aesar
A13922.18 |
50 g |
30.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11401648
|
Thermo Scientific Alfa Aesar
A13922.30 |
250 g |
112.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11481638
|
Thermo Scientific Alfa Aesar
A13922.0B |
1000 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
626-43-7 | |
162.013 | |
UQRLKWGPEVNVHT-UHFFFAOYSA-N | |
12281 | |
3,5-dichloroaniline |
C6H5Cl2N | |
MFCD00007774 | |
benzenamine, 3,5-dichloro, m-dichloroaniline, aniline, 3,5-dichloro, 3,5-dichlorobenzenamine, 3,5-dichloro benzenamine, unii-oz75zm1s3g, 3,5-dichloranilin, 3,5 dichloraniline, ccris 2396, 3,5-dichloro aniline | |
CHEBI:19904 | |
C1=C(C=C(C=C1Cl)Cl)N |
Specifications
626-43-7 | |
1.58 | |
133°C (271°F) | |
MFCD00007774 | |
636492 | |
UQRLKWGPEVNVHT-UHFFFAOYSA-N | |
3,5-dichloroaniline | |
12281 | |
162.02 | |
3,5-Dichloroaniline |
49°C to 53°C | |
259°C to 260°C (741 mmHg) | |
C6H5Cl2N | |
UN3442 | |
benzenamine, 3,5-dichloro, m-dichloroaniline, aniline, 3,5-dichloro, 3,5-dichlorobenzenamine, 3,5-dichloro benzenamine, unii-oz75zm1s3g, 3,5-dichloranilin, 3,5 dichloraniline, ccris 2396, 3,5-dichloro aniline | |
C1=C(C=C(C=C1Cl)Cl)N | |
162.013 | |
CHEBI:19904 | |
98% |
Safety and Handling
GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
H301+H311+H331-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
missing translation for 'einecsNumber' : 210-948-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only