missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzoic acid, 98%, Thermo Scientific™

Product Code. 11477987
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Dibromobenzoic acid may be used for the syntheses (+)-menthyl 3,5-dibromobenzoate, di-tert-butyl 4-[2-(tert-butoxycarbonyl)ethyl]-4-(3,5-dibromobenzamido)heptanedioate2, (L)-methyl 2-(3,5-dibromobenzamido)-3-phenylpropanoate.

Solubility
Very faint turbidity in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-58-6
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00051758
InChI Key SFTFNJZWZHASAQ-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-dibromo, 3,5-dibromobenzoicacid, benzoicacid, 3,5-dibromo, buttpark 99\57-19, pubchem3861, acmc-209mwz, maybridge1_003318, 3,5-dibromo-benzoic acid, 4-09-00-01028 beilstein handbook reference, ksc358a3h
PubChem CID 12063
IUPAC Name 3,5-dibromobenzoic acid
SMILES OC(=O)C1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 216°C to 224°C
Quantity 25 g
Beilstein 1940691
Solubility Information Very faint turbidity in methanol.
Formula Weight 279.93
Percent Purity 98%
Chemical Name or Material 3,5-Dibromobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'