missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diamino-1,2,4-triazole, 98%, Thermo Scientific Chemicals

Product Code. 10511252
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1455-77-2
Molecular Formula C2H5N5
Molecular Weight (g/mol) 99.09
MDL Number MFCD00005233
InChI Key PKWIYNIDEDLDCJ-UHFFFAOYSA-N
Synonym guanazole, 3,5-diamino-1,2,4-triazole, 4h-1,2,4-triazole-3,5-diamine, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, s-triazole, 3,5-diamino, 1,2,4-triazolidine, 3,5-diimino, unii-i01twm5267, ccris 3745, 3-amino-1h-1,2,4-triazol-5-ylamine
PubChem CID 15078
ChEBI CHEBI:75425
IUPAC Name 1H-1,2,4-triazole-3,5-diamine
SMILES C1(=NC(=NN1)N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 204°C to 208°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 26, 193
Formula Weight 99.09
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3,5-Diamino-1,2,4-triazole
Show More Show Less
Product Identifier
  • 3,5-Diamino-1,2,4-triazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'