missing translation for 'onlineSavingsMsg'
Learn More

3,5,5-Trimethylcyclohexane-1,2-dione, 99%, Thermo Scientific™

Product Code. 11405237
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57696-89-6
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
MDL Number MFCD02181023
InChI Key WEUZZYGESXCTCU-UHFFFAOYSA-N
Synonym 3,5,5-trimethyl-1,2-cyclohexanedione, 1,2-cyclohexanedione, 3,5,5-trimethyl, 3,5,5-trimethyl-1.2-cyclohexanedione, acmc-1aooj, ksc274g5l, 3,5,5-trimethyl-1,2 cyclohexanedione, 3,5,5-trimethyl cyclohexane-1,2-dione, 3,5,5-trimethylcyclohexane-1,2-dione 5g
PubChem CID 93772
IUPAC Name 3,5,5-trimethylcyclohexane-1,2-dione
SMILES CC1CC(CC(=O)C1=O)(C)C

Specifications

Melting Point 89°C to 92°C
Flash Point >100°C (212°F)
Quantity 25 g
Formula Weight 154.21
Percent Purity 99%
Chemical Name or Material 3,5,5-Trimethylcyclohexane-1,2-dione
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.