missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Nitrophenyl)-1H-pyrazole, 97%, Thermo Scientific Chemicals

Product Code. 15495047
Click to view available options
Quantity:
500 mg
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20583-31-7
Molecular Formula C9H7N3O2
Molecular Weight (g/mol) 189.17
MDL Number MFCD00665859
InChI Key IPIYADCDDIUVPS-UHFFFAOYSA-N
Synonym 3-4-nitrophenyl-1h-pyrazole, 3-4-nitrophenyl pyrazole, 5-4-nitrophenyl-1h-pyrazole, 3-4-nitro-phenyl-1h-pyrazole, 5-4-nitrophenyl pyrazole, 1h-pyrazole, 3-4-nitrophenyl, 5-4-nitro-phenyl-1h-pyrazole, 3-4-nitrophenyl-2h-pyrazole, nitrophenylpyrazole, p-nitrophenylpyrazole
PubChem CID 2737071
IUPAC Name 5-(4-nitrophenyl)-1H-pyrazole
SMILES [O-][N+](=O)C1=CC=C(C=C1)C1=CC=NN1

Specifications

Melting Point 194°C to 195°C
Quantity 500 mg
Formula Weight 189.18
Percent Purity 97%
Chemical Name or Material 3-(4-Nitrophenyl)-1H-pyrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'