missing translation for 'onlineSavingsMsg'
Learn More

3-[4-Methoxy-3-(trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™

43.00 € - 152.00 €

Chemical Identifiers

CAS 1005205-20-8
Molecular Formula C11H11F3O3
Molecular Weight (g/mol) 248.201
MDL Number MFCD11519317
InChI Key BZGRRQPEHHXJNW-UHFFFAOYSA-N
Synonym 3-4-methoxy-3-trifluoromethyl phenyl propionic acid, 3-4-methoxy-3-trifluoromethyl phenyl propanoic acid
PubChem CID 57363570
IUPAC Name 3-[4-methoxy-3-(trifluoromethyl)phenyl]propanoic acid
SMILES COC1=C(C=C(C=C1)CCC(=O)O)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15478085
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32428.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15468085
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32428.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1005205-20-8
248.201
BZGRRQPEHHXJNW-UHFFFAOYSA-N
57363570
COC1=C(C=C(C=C1)CCC(=O)O)C(F)(F)F
C11H11F3O3
MFCD11519317
3-4-methoxy-3-trifluoromethyl phenyl propionic acid, 3-4-methoxy-3-trifluoromethyl phenyl propanoic acid
3-[4-methoxy-3-(trifluoromethyl)phenyl]propanoic acid

Specifications

1005205-20-8
C11H11F3O3
3-4-methoxy-3-trifluoromethyl phenyl propionic acid, 3-4-methoxy-3-trifluoromethyl phenyl propanoic acid
COC1=C(C=C(C=C1)CCC(=O)O)C(F)(F)F
248.201
248.2
3-[4-Methoxy-3-(trifluoromethyl)phenyl]propionic acid
94°C to 97°C
MFCD11519317
BZGRRQPEHHXJNW-UHFFFAOYSA-N
3-[4-methoxy-3-(trifluoromethyl)phenyl]propanoic acid
57363570
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications