missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethylthieno[2,3-b]thiophene-2-carbaldehyde, 97%, Thermo Scientific™
66.00 € - 214.00 €
Chemical Identifiers
CAS | 159709-36-1 |
---|---|
Molecular Formula | C9H8OS2 |
Molecular Weight (g/mol) | 196.282 |
MDL Number | MFCD00052282 |
InChI Key | WEAXSPMHCXWYPF-UHFFFAOYSA-N |
Synonym | 3,4-dimethylthieno 2,3-b thiophene-2-carbaldehyde, thieno 2,3-b thiophene-2-carboxaldehyde,3,4-dimethyl, 3,4-dimethyl-2-formylthieno 2,3-b thiophene, 3,4-dimethylthieno 2,3-b thiophene-5-carbaldehyde, 3,4-dimethylthieno 2,3-b thiophene-2-carboxaldehyde, 3,4-dimethylthiopheno 2,3-b thiophene-2-carbaldehyde |
PubChem CID | 2778771 |
IUPAC Name | 3,4-dimethylthieno[2,3-b]thiophene-5-carbaldehyde |
SMILES | CC1=CSC2=C1C(=C(S2)C=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10691673
|
Thermo Scientific Maybridge
KM03483DA |
1 g |
66.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10488494
|
Thermo Scientific Maybridge
KM03483DE |
5 g |
214.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
159709-36-1 | |
196.282 | |
WEAXSPMHCXWYPF-UHFFFAOYSA-N | |
2778771 | |
CC1=CSC2=C1C(=C(S2)C=O)C |
C9H8OS2 | |
MFCD00052282 | |
3,4-dimethylthieno 2,3-b thiophene-2-carbaldehyde, thieno 2,3-b thiophene-2-carboxaldehyde,3,4-dimethyl, 3,4-dimethyl-2-formylthieno 2,3-b thiophene, 3,4-dimethylthieno 2,3-b thiophene-5-carbaldehyde, 3,4-dimethylthieno 2,3-b thiophene-2-carboxaldehyde, 3,4-dimethylthiopheno 2,3-b thiophene-2-carbaldehyde | |
3,4-dimethylthieno[2,3-b]thiophene-5-carbaldehyde |
Specifications
159709-36-1 | |
97% | |
C9H8OS2 | |
3,4-dimethylthieno 2,3-b thiophene-2-carbaldehyde, thieno 2,3-b thiophene-2-carboxaldehyde,3,4-dimethyl, 3,4-dimethyl-2-formylthieno 2,3-b thiophene, 3,4-dimethylthieno 2,3-b thiophene-5-carbaldehyde, 3,4-dimethylthieno 2,3-b thiophene-2-carboxaldehyde, 3,4-dimethylthiopheno 2,3-b thiophene-2-carbaldehyde | |
CC1=CSC2=C1C(=C(S2)C=O)C | |
196.282 | |
196.29 | |
3, 4-Dimethylthieno[2, 3-b]thiophene-2-carbaldehyde |
97.0 | |
Amber glass bottle | |
MFCD00052282 | |
WEAXSPMHCXWYPF-UHFFFAOYSA-N | |
3,4-dimethylthieno[2,3-b]thiophene-5-carbaldehyde | |
2778771 | |
97% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title