missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethoxyphenethyl alcohol, 98%, Thermo Scientific Chemicals
84.80 € - 235.00 €
Chemical Identifiers
CAS | 7417-21-2 |
---|---|
Molecular Formula | C10H14O3 |
Molecular Weight (g/mol) | 182.22 |
MDL Number | MFCD00002894 |
InChI Key | SRQAJMUHZROVHW-UHFFFAOYSA-N |
Synonym | 3,4-dimethoxyphenethyl alcohol, 2-3,4-dimethoxyphenyl ethanol, 3,4-dimethoxyphenethylalcohol, 2-3,4-dimethoxyphenyl ethan-1-ol, 3,4-dimethoxyphenethanol, homoveratryl alcohol, benzeneethanol, 3,4-dimethoxy, 3,4-dimethoxybenzeneethanol, 3,4-dimethoxy-.beta.-phenethyl alcohol, 1-2-hydroxyethyl-3,4-dimethoxybenzene |
PubChem CID | 81911 |
IUPAC Name | 2-(3,4-dimethoxyphenyl)ethanol |
SMILES | COC1=CC=C(CCO)C=C1OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10562401
|
Thermo Scientific Acros
189000100 |
10 g | Glass bottle |
84.80 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10591564
|
Thermo Scientific Acros
189000500 |
50 g | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7417-21-2 | |
182.22 | |
SRQAJMUHZROVHW-UHFFFAOYSA-N | |
81911 | |
COC1=CC=C(CCO)C=C1OC |
C10H14O3 | |
MFCD00002894 | |
3,4-dimethoxyphenethyl alcohol, 2-3,4-dimethoxyphenyl ethanol, 3,4-dimethoxyphenethylalcohol, 2-3,4-dimethoxyphenyl ethan-1-ol, 3,4-dimethoxyphenethanol, homoveratryl alcohol, benzeneethanol, 3,4-dimethoxy, 3,4-dimethoxybenzeneethanol, 3,4-dimethoxy-.beta.-phenethyl alcohol, 1-2-hydroxyethyl-3,4-dimethoxybenzene | |
2-(3,4-dimethoxyphenyl)ethanol |
Specifications
7417-21-2 | |
100.0 | |
Yellow | |
>110°C | |
97.5% min. (GC) | |
(CH3O)2C6H3CH2CH2OH | |
06, III, 6333 | |
SRQAJMUHZROVHW-UHFFFAOYSA-N | |
2-(3,4-dimethoxyphenyl)ethanol | |
81911 | |
98% | |
3,4-Dimethoxyphenethyl alcohol |
97.5 | |
44°C to 48°C | |
172°C to 174°C (17.0 mmHg) | |
Authentic | |
C10H14O3 | |
MFCD00002894 | |
3,4-dimethoxyphenethyl alcohol, 2-3,4-dimethoxyphenyl ethanol, 3,4-dimethoxyphenethylalcohol, 2-3,4-dimethoxyphenyl ethan-1-ol, 3,4-dimethoxyphenethanol, homoveratryl alcohol, benzeneethanol, 3,4-dimethoxy, 3,4-dimethoxybenzeneethanol, 3,4-dimethoxy-.beta.-phenethyl alcohol, 1-2-hydroxyethyl-3,4-dimethoxybenzene | |
COC1=CC=C(CCO)C=C1OC | |
182.22 | |
182.22 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 231-032-5
Product Identifier
- 3,4-Dimethoxyphenethyl alcohol
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title