missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzoyl chloride, 98%, Thermo Scientific Chemicals

46.43 € - 152.76 €

Chemical Identifiers

CAS 3535-37-3
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.62
MDL Number MFCD00000674
InChI Key VIOBGCWEHLRBEP-UHFFFAOYSA-N
Synonym veratroyl chloride, 3,4-dimethoxybenzoylchloride, 3,4-dimethoxybenzene-1-carbonyl chloride, benzoyl chloride, 3,4-dimethoxy, 3,4-dimethoxy benzoyl chloride, 3,4-dimethoxy-benzoyl chloride, 3,4-dimethoxy-1-benzenecarbonyl chloride, veratroylchlorid, zlchem 1258, ksc225m7t
PubChem CID 77070
IUPAC Name 3,4-dimethoxybenzoyl chloride
SMILES COC1=C(C=C(C=C1)C(=O)Cl)OC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10668211
View Documents
Thermo Scientific Acros
305600050
5 g Glass bottle
46.43 €
5g
Please to purchase this item. Need a web account? Register with us today!
10239553
View Documents
Thermo Scientific Acros
305600250
25 g Glass bottle
152.76 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3535-37-3
200.62
VIOBGCWEHLRBEP-UHFFFAOYSA-N
77070
COC1=C(C=C(C=C1)C(=O)Cl)OC
C9H9ClO3
MFCD00000674
veratroyl chloride, 3,4-dimethoxybenzoylchloride, 3,4-dimethoxybenzene-1-carbonyl chloride, benzoyl chloride, 3,4-dimethoxy, 3,4-dimethoxy benzoyl chloride, 3,4-dimethoxy-benzoyl chloride, 3,4-dimethoxy-1-benzenecarbonyl chloride, veratroylchlorid, zlchem 1258, ksc225m7t
3,4-dimethoxybenzoyl chloride

Specifications

3535-37-3
100.0
White to Gray
97.5% min. (GC)
(CH3O)2C6H3COCl
veratroyl chloride, 3,4-dimethoxybenzoylchloride, 3,4-dimethoxybenzene-1-carbonyl chloride, benzoyl chloride, 3,4-dimethoxy, 3,4-dimethoxy benzoyl chloride, 3,4-dimethoxy-benzoyl chloride, 3,4-dimethoxy-1-benzenecarbonyl chloride, veratroylchlorid, zlchem 1258, ksc225m7t
COC1=C(C=C(C=C1)C(=O)Cl)OC
200.62
200.62
Crystals, Flakes or Chunks
97.5
68.0°C to 72.0°C
Authentic
C9H9ClO3
MFCD00000674
VIOBGCWEHLRBEP-UHFFFAOYSA-N
3,4-dimethoxybenzoyl chloride
77070
98%
3, 4-Dimethoxybenzoyl chloride, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 222-568-0

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.