missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethoxyacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 10612413
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1131-62-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00008737
InChI Key IQZLUWLMQNGTIW-UHFFFAOYSA-N
Synonym 3',4'-dimethoxyacetophenone, 1-3,4-dimethoxyphenyl ethanone, 3,4-dimethoxyacetophenone, acetoveratrone, ethanone, 1-3,4-dimethoxyphenyl, 1-3,4-dimethoxyphenyl ethan-1-one, 3,4-dimethoxyphenyl methyl ketone, unii-5rv6436s8a, acetophenone, 3',4'-dimethoxy, acetophenone,4'-dimethoxy
PubChem CID 14328
ChEBI CHEBI:86576
IUPAC Name 1-(3,4-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OC)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48.0°C to 52.0°C
Color Yellow to Beige
Boiling Point 286.0°C to 288.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3COCH3
Quantity 5 g
Beilstein 08, II, 298
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol
Formula Weight 180.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3', 4'-Dimethoxyacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 3',4'-Dimethoxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.