missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dihydroxypropiophenone, 98%, Thermo Scientific Chemicals

Product Code. 11415503
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7451-98-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00016440
InChI Key HNWIHBDMOYWCGX-UHFFFAOYSA-N
Synonym 3',4'-dihydroxypropiophenone, 3,4-dihydroxypropiophenone, propiophenone, 3',4'-dihydroxy, 1-3,4-dihydroxyphenyl propan-1-one, 1-propanone, 1-3,4-dihydroxyphenyl, 1-3,4-dihydroxyphenyl-1-propanone, 1-propanone,1-3,4-dihydroxyphenyl, 4-propionylbrenzcatechin, propiophenone,4'-dihydroxy, 3', 4'-dihydroxypropiophenone
PubChem CID 96267
IUPAC Name 1-(3,4-dihydroxyphenyl)propan-1-one
SMILES CCC(=O)C1=CC(=C(C=C1)O)O

Specifications

Melting Point 145°C to 146°C
Quantity 1 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 3',4'-Dihydroxypropiophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'