Learn More
3,4-Dihydroxybenzaldoxime, 98%, Thermo Scientific™
50.80 € - 178.00 €
Chemical Identifiers
CAS | 3343-59-7 |
---|---|
Molecular Formula | C7H7NO3 |
Molecular Weight (g/mol) | 153.137 |
MDL Number | MFCD01070622 |
InChI Key | XQTLVFXPCMGDOB-UHFFFAOYSA-N |
Synonym | 3,4-dihydroxybenzaldoxime, 3,4-dihydroxybenzaldehyde oxime, acmc-20anf4, 4-hydroxyimino methyl benzene-1,2-diol, 3,4-dihydroxybenzaldehydeoxime, nufmsecagoxhav-uhfffaoysa-n, benzaldehyde,3,4-dihydroxy-, oxime, 4-1e-hydroxyimino methyl benzene-1,2-diol, 2-hydroxy-4-hydroxyamino methylidene cyclohexa-2,5-dien-1-one |
PubChem CID | 6798780 |
IUPAC Name | 2-hydroxy-4-[(hydroxyamino)methylidene]cyclohexa-2,5-dien-1-one |
SMILES | C1=CC(=O)C(=CC1=CNO)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11314755
|
Thermo Scientific Alfa Aesar
L17766.06 |
5 g |
50.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324755
|
Thermo Scientific Alfa Aesar
L17766.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
3343-59-7 | |
153.137 | |
XQTLVFXPCMGDOB-UHFFFAOYSA-N | |
6798780 | |
C1=CC(=O)C(=CC1=CNO)O |
C7H7NO3 | |
MFCD01070622 | |
3,4-dihydroxybenzaldoxime, 3,4-dihydroxybenzaldehyde oxime, acmc-20anf4, 4-hydroxyimino methyl benzene-1,2-diol, 3,4-dihydroxybenzaldehydeoxime, nufmsecagoxhav-uhfffaoysa-n, benzaldehyde,3,4-dihydroxy-, oxime, 4-1e-hydroxyimino methyl benzene-1,2-diol, 2-hydroxy-4-hydroxyamino methylidene cyclohexa-2,5-dien-1-one | |
2-hydroxy-4-[(hydroxyamino)methylidene]cyclohexa-2,5-dien-1-one |
Specifications
3343-59-7 | |
C7H7NO3 | |
3126402 | |
XQTLVFXPCMGDOB-UHFFFAOYSA-N | |
2-hydroxy-4-[(hydroxyamino)methylidene]cyclohexa-2,5-dien-1-one | |
6798780 | |
98% |
155°C to 157°C (decomposition) | |
MFCD01070622 | |
3,4-dihydroxybenzaldoxime, 3,4-dihydroxybenzaldehyde oxime, acmc-20anf4, 4-hydroxyimino methyl benzene-1,2-diol, 3,4-dihydroxybenzaldehydeoxime, nufmsecagoxhav-uhfffaoysa-n, benzaldehyde,3,4-dihydroxy-, oxime, 4-1e-hydroxyimino methyl benzene-1,2-diol, 2-hydroxy-4-hydroxyamino methylidene cyclohexa-2,5-dien-1-one | |
C1=CC(=O)C(=CC1=CNO)O | |
153.137 | |
153.14 | |
3,4-Dihydroxybenzaldoxime |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.