Learn More
(3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol, 97%, Thermo Scientific™
37.00 € - 323.00 €
Chemical Identifiers
CAS | 304669-35-0 |
---|---|
Molecular Formula | C7H14O2Si |
Molecular Weight (g/mol) | 158.272 |
MDL Number | MFCD09265116 |
InChI Key | ZMDNGTLKIVNNCF-UHFFFAOYSA-N |
Synonym | 3,4-dihydro-2h-pyran-6-yl dimethylsilanol, 5,6-dihydro-4h-pyran-2-yl dimethylsilanol, 2-5,6-dihydro-4h-pyranyl dimethylsilanol, silanol, 3,4-dihydro-2h-pyran-6-yl dimethyl, 5,6-dihydro-4h-pyran-2-yldimethylsilanol, amtsi069 |
PubChem CID | 11332616 |
IUPAC Name | 3,4-dihydro-2H-pyran-6-yl-hydroxy-dimethylsilane |
SMILES | C[Si](C)(C1=CCCCO1)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
304669-35-0 | |
158.272 | |
ZMDNGTLKIVNNCF-UHFFFAOYSA-N | |
11332616 | |
C[Si](C)(C1=CCCCO1)O |
C7H14O2Si | |
MFCD09265116 | |
3,4-dihydro-2h-pyran-6-yl dimethylsilanol, 5,6-dihydro-4h-pyran-2-yl dimethylsilanol, 2-5,6-dihydro-4h-pyranyl dimethylsilanol, silanol, 3,4-dihydro-2h-pyran-6-yl dimethyl, 5,6-dihydro-4h-pyran-2-yldimethylsilanol, amtsi069 | |
3,4-dihydro-2H-pyran-6-yl-hydroxy-dimethylsilane |
Specifications
54°C (0.8mmHg) | |
C7H14O2Si | |
91°C (196°F) | |
ZMDNGTLKIVNNCF-UHFFFAOYSA-N | |
3,4-dihydro-2H-pyran-6-yl-hydroxy-dimethylsilane | |
11332616 | |
97% | |
1.005 |
304669-35-0 | |
MFCD09265116 | |
3,4-dihydro-2h-pyran-6-yl dimethylsilanol, 5,6-dihydro-4h-pyran-2-yl dimethylsilanol, 2-5,6-dihydro-4h-pyranyl dimethylsilanol, silanol, 3,4-dihydro-2h-pyran-6-yl dimethyl, 5,6-dihydro-4h-pyran-2-yldimethylsilanol, amtsi069 | |
C[Si](C)(C1=CCCCO1)O | |
158.272 | |
158.27 | |
1.475 | |
(3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Nitrogen
Your input is important to us. Please complete this form to provide feedback related to the content on this product.