missing translation for 'onlineSavingsMsg'
Learn More

3,4'-Dichloropropiophenone, 97%, Thermo Scientific Chemicals

55.88 € - 198.99 €

Chemical Identifiers

CAS 3946-29-0
Molecular Formula C9H8Cl2O
Molecular Weight (g/mol) 203.06
MDL Number MFCD00000992
InChI Key AYFJBHFMQODYBC-UHFFFAOYSA-N
Synonym 3,4'-dichloropropiophenone, 3-chloro-1-4-chlorophenyl propan-1-one, 1-propanone, 3-chloro-1-4-chlorophenyl, 2-chloroethyl-4-chlorophenylketone, pubchem13270, acmc-209j5n, beta,4-dichloropropiophenone, 4',beta-dichloropropiophenone, 3-chloro-4'-chloropropiophenone, attercop-chm at130375
PubChem CID 77550
IUPAC Name 3-chloro-1-(4-chlorophenyl)propan-1-one
SMILES ClCCC(=O)C1=CC=C(Cl)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11460903
View Documents
Thermo Scientific Alfa Aesar
A13311.09
10 g
55.88 €
10g
Please to purchase this item. Need a web account? Register with us today!
11470903
View Documents
Thermo Scientific Alfa Aesar
A13311.18
50 g
198.99 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3946-29-0
203.06
AYFJBHFMQODYBC-UHFFFAOYSA-N
77550
ClCCC(=O)C1=CC=C(Cl)C=C1
C9H8Cl2O
MFCD00000992
3,4'-dichloropropiophenone, 3-chloro-1-4-chlorophenyl propan-1-one, 1-propanone, 3-chloro-1-4-chlorophenyl, 2-chloroethyl-4-chlorophenylketone, pubchem13270, acmc-209j5n, beta,4-dichloropropiophenone, 4',beta-dichloropropiophenone, 3-chloro-4'-chloropropiophenone, attercop-chm at130375
3-chloro-1-(4-chlorophenyl)propan-1-one

Specifications

3946-29-0
135°C to 137°C (0.6 mmHg)
C9H8Cl2O
1866915
AYFJBHFMQODYBC-UHFFFAOYSA-N
3-chloro-1-(4-chlorophenyl)propan-1-one
77550
97%
48°C to 52°C
>110°C (230°F)
MFCD00000992
3,4'-dichloropropiophenone, 3-chloro-1-4-chlorophenyl propan-1-one, 1-propanone, 3-chloro-1-4-chlorophenyl, 2-chloroethyl-4-chlorophenylketone, pubchem13270, acmc-209j5n, beta,4-dichloropropiophenone, 4',beta-dichloropropiophenone, 3-chloro-4'-chloropropiophenone, attercop-chm at130375
ClCCC(=O)C1=CC=C(Cl)C=C1
203.06
203.07
3,4'-Dichloropropiophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.