Learn More
3,4-Dichlorophenylacetic acid, 98%, Thermo Scientific Chemicals
26.50 € - 249.00 €
Chemical Identifiers
CAS | 5807-30-7 |
---|---|
Molecular Formula | C8H6Cl2O2 |
Molecular Weight (g/mol) | 205.034 |
MDL Number | MFCD00004333 |
InChI Key | ZOUPGSMSNQLUNW-UHFFFAOYSA-N |
Synonym | 3,4-dichlorophenylacetic acid, 2-3,4-dichlorophenyl acetic acid, 3,4-dichlorophenyl acetic acid, 3,4-dichlorobenzene acetic acid, benzeneacetic acid, 3,4-dichloro, pubchem20195, acmc-209m2g, 3,4dichlorophenylacetic acid, 3,4-dichlorophenylaceticacid |
PubChem CID | 79874 |
IUPAC Name | 2-(3,4-dichlorophenyl)acetic acid |
SMILES | C1=CC(=C(C=C1CC(=O)O)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11467567
|
Thermo Scientific Alfa Aesar
A15653.06 |
5 g |
26.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11477567
|
Thermo Scientific Alfa Aesar
A15653.14 |
25 g |
75.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11487567
|
Thermo Scientific Alfa Aesar
A15653.22 |
100 g |
249.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in the parallel solid phase synthesis of tetra substituted diethylenetriamines via selective amide alkylation and exhaustive reduction of N-acylated dipeptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the parallel solid phase synthesis of tetra substituted diethylenetriamines via selective amide alkylation and exhaustive reduction of N-acylated dipeptides.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
5807-30-7 | |
205.034 | |
ZOUPGSMSNQLUNW-UHFFFAOYSA-N | |
79874 | |
C1=CC(=C(C=C1CC(=O)O)Cl)Cl |
C8H6Cl2O2 | |
MFCD00004333 | |
3,4-dichlorophenylacetic acid, 2-3,4-dichlorophenyl acetic acid, 3,4-dichlorophenyl acetic acid, 3,4-dichlorobenzene acetic acid, benzeneacetic acid, 3,4-dichloro, pubchem20195, acmc-209m2g, 3,4dichlorophenylacetic acid, 3,4-dichlorophenylaceticacid | |
2-(3,4-dichlorophenyl)acetic acid |
Specifications
5807-30-7 | |
Odorless | |
MFCD00004333 | |
3,4-dichlorophenylacetic acid, 2-3,4-dichlorophenyl acetic acid, 3,4-dichlorophenyl acetic acid, 3,4-dichlorobenzene acetic acid, benzeneacetic acid, 3,4-dichloro, pubchem20195, acmc-209m2g, 3,4dichlorophenylacetic acid, 3,4-dichlorophenylaceticacid | |
ZOUPGSMSNQLUNW-UHFFFAOYSA-N | |
2-(3,4-dichlorophenyl)acetic acid | |
79874 | |
98% |
87°C to 92°C | |
C8H6Cl2O2 | |
1953781 | |
Insoluble in water. | |
C1=CC(=C(C=C1CC(=O)O)Cl)Cl | |
205.034 | |
205.04 | |
3,4-Dichlorophenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 227-368-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only