missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorobenzoyl)propionic acid, 98%, Thermo Scientific™

Product Code. 11449306
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3984-34-7
Molecular Formula C10H9ClO3
Molecular Weight (g/mol) 212.63
MDL Number MFCD00002794
InChI Key AHVASTJJVAYFPY-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl-4-oxobutanoic acid, 3-4-chlorobenzoyl propionic acid, 4-4-chloro-phenyl-4-oxo-butyric acid, 3-4-chlorobenzoyl propanoic acid, 3-p-chlorobenzoyl propionic acid, 4-4-chlorophenyl-4-oxobutanoicacid, maybridge1_006214, ncistruc1_000312, ncistruc2_000239, asischem d51605
PubChem CID 77604
IUPAC Name 4-(4-chlorophenyl)-4-oxobutanoic acid
SMILES OC(=O)CCC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 128°C to 130°C
Quantity 25 g
Beilstein 2104409
Formula Weight 212.63
Percent Purity 98%
Chemical Name or Material 3-(4-Chlorobenzoyl)propionic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.