missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,9,10-Perylenetetracarboxylic dianhydride, 98%, Thermo Scientific Chemicals
39.10 € - 95.70 €
Chemical Identifiers
CAS | 128-69-8 |
---|---|
Molecular Formula | C24H8O6 |
Molecular Weight (g/mol) | 392.32 |
MDL Number | MFCD00006916 |
InChI Key | CLYVDMAATCIVBF-UHFFFAOYSA-N |
Synonym | 3,4,9,10-perylenetetracarboxylic dianhydride, pigment red 224, ptcda, perylene-3,4,9,10-tetracarboxylic dianhydride, perylenetetracarboxylic anhydride, anthra 2,1,9-def:6,5,10-d'e'f' diisochromene-1,3,8,10-tetraone, perylo 3,4-cd:9,10-c'd' dipyran-1,3,8,10-tetrone, perylenetetracarboxylic acid dianhydride, 3,4:9,10-perylenetetracarboxylic anhydride |
PubChem CID | 67191 |
IUPAC Name | 7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone |
SMILES | O=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10246923
|
Thermo Scientific Acros
130080250 |
25 g | Glass bottle |
39.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10666712
|
Thermo Scientific Acros
130081000 |
100 g | Glass bottle |
95.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
128-69-8 | |
392.32 | |
CLYVDMAATCIVBF-UHFFFAOYSA-N | |
67191 | |
O=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56 |
C24H8O6 | |
MFCD00006916 | |
3,4,9,10-perylenetetracarboxylic dianhydride, pigment red 224, ptcda, perylene-3,4,9,10-tetracarboxylic dianhydride, perylenetetracarboxylic anhydride, anthra 2,1,9-def:6,5,10-d'e'f' diisochromene-1,3,8,10-tetraone, perylo 3,4-cd:9,10-c'd' dipyran-1,3,8,10-tetrone, perylenetetracarboxylic acid dianhydride, 3,4:9,10-perylenetetracarboxylic anhydride | |
7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone |
Specifications
128-69-8 | |
100.0 | |
Red | |
97.5% min | |
MFCD00006916 | |
Solubility in water: insoluble. Other solubilities: sparingly soluble in dil.alkalies and conc. h2so4, insoluble in most common organic solvents | |
O=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56 | |
392.32 | |
392.32 | |
Powder |
97.5 | |
>350.0°C | |
Authentic | |
C24H8O6 | |
3,4,9,10-perylenetetracarboxylic dianhydride, pigment red 224, ptcda, perylene-3,4,9,10-tetracarboxylic dianhydride, perylenetetracarboxylic anhydride, anthra 2,1,9-def:6,5,10-d'e'f' diisochromene-1,3,8,10-tetraone, perylo 3,4-cd:9,10-c'd' dipyran-1,3,8,10-tetrone, perylenetetracarboxylic acid dianhydride, 3,4:9,10-perylenetetracarboxylic anhydride | |
CLYVDMAATCIVBF-UHFFFAOYSA-N | |
7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone | |
67191 | |
98% | |
3,4,9,10-Perylenetetracarboxylic dianhydride, 98% |
Safety and Handling
MOISTURE SENSITIVE
missing translation for 'einecsNumber' : 204-905-3
missing translation for 'tsca' : TSCA
RUO – Research Use Only