Learn More
(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 98+%, Thermo Scientific Chemicals
149.00 € - 585.00 €
Chemical Identifiers
CAS | 20007-72-1 |
---|---|
Molecular Formula | C11H14O2 |
Molecular Weight (g/mol) | 178.23 |
MDL Number | MFCD00001687 |
InChI Key | DNHDRUMZDHWHKG-UHFFFAOYNA-N |
Synonym | wieland-miescher ketone, 9-methyl-delta-5 10-octaline-1,6-dione, 9-methyl-5 10-octaline-1,6-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl, 8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione, wieland-michler ketone, wieland-miescher-keton, miescher-wieland ketone, wieland cmiescher ketone, 9-s-methyl-.delta.-5 10-octalin-1,6-dione |
PubChem CID | 89262 |
IUPAC Name | 8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione |
SMILES | CC12CCC(=O)C=C1CCCC2=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15460827
|
Thermo Scientific Alfa Aesar
L13326.03 |
1 g |
149.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15470827
|
Thermo Scientific Alfa Aesar
L13326.06 |
5 g |
585.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20007-72-1 | |
178.23 | |
DNHDRUMZDHWHKG-UHFFFAOYNA-N | |
89262 | |
CC12CCC(=O)C=C1CCCC2=O |
C11H14O2 | |
MFCD00001687 | |
wieland-miescher ketone, 9-methyl-delta-5 10-octaline-1,6-dione, 9-methyl-5 10-octaline-1,6-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl, 8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione, wieland-michler ketone, wieland-miescher-keton, miescher-wieland ketone, wieland cmiescher ketone, 9-s-methyl-.delta.-5 10-octalin-1,6-dione | |
8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione |
Specifications
20007-72-1 | |
C11H14O2 | |
3201286 | |
DNHDRUMZDHWHKG-UHFFFAOYNA-N | |
8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione | |
89262 | |
≥98% |
47°C to 50°C | |
MFCD00001687 | |
wieland-miescher ketone, 9-methyl-delta-5 10-octaline-1,6-dione, 9-methyl-5 10-octaline-1,6-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl, 8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione, wieland-michler ketone, wieland-miescher-keton, miescher-wieland ketone, wieland cmiescher ketone, 9-s-methyl-.delta.-5 10-octalin-1,6-dione | |
CC12CCC(=O)C=C1CCCC2=O | |
178.23 | |
178.23 | |
(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione |
Safety and Handling
missing translation for 'einecsNumber' : 243-463-6
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.