Learn More
3-(4,5-Diphenyl-1,3-oxazol-2-yl)propanoic acid, 97%, Thermo Scientific™
25.80 € - 99.80 €
Chemical Identifiers
CAS | 21256-18-8 |
---|---|
Molecular Formula | C18H15NO3 |
Molecular Weight (g/mol) | 293.32 |
MDL Number | MFCD00215977 |
InChI Key | OFPXSFXSNFPTHF-UHFFFAOYSA-N |
Synonym | oxaprozin, daypro, alvo, 3-4,5-diphenyl-1,3-oxazol-2-yl propanoic acid, oxaprozine, 3-4,5-diphenyloxazol-2-yl propanoic acid, deflam, oxaprozina, oxaprozinum, voir |
PubChem CID | 4614 |
ChEBI | CHEBI:7822 |
IUPAC Name | 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid |
SMILES | OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10742133
|
Thermo Scientific Maybridge
SS00042DA |
1 g |
25.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10703377
|
Thermo Scientific Maybridge
SS00042DE |
5 g |
99.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
21256-18-8 | |
293.32 | |
OFPXSFXSNFPTHF-UHFFFAOYSA-N | |
4614 | |
3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid |
C18H15NO3 | |
MFCD00215977 | |
oxaprozin, daypro, alvo, 3-4,5-diphenyl-1,3-oxazol-2-yl propanoic acid, oxaprozine, 3-4,5-diphenyloxazol-2-yl propanoic acid, deflam, oxaprozina, oxaprozinum, voir | |
CHEBI:7822 | |
OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
21256-18-8 | |
97% | |
C18H15NO3 | |
15, 7023 | |
OFPXSFXSNFPTHF-UHFFFAOYSA-N | |
3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid | |
4614 | |
293.32 | |
3-(4, 5-Diphenyl-1, 3-oxazol-2-yl)propanoic acid |
97.0 | |
Amber glass bottle | |
MFCD00215977 | |
oxaprozin, daypro, alvo, 3-4,5-diphenyl-1,3-oxazol-2-yl propanoic acid, oxaprozine, 3-4,5-diphenyloxazol-2-yl propanoic acid, deflam, oxaprozina, oxaprozinum, voir | |
OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1 | |
293.32 | |
CHEBI:7822 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.