missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Piperidinyloxy)benzonitrile, 99%, Thermo Scientific Chemicals

Product Code. 11419680
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902836-93-5
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
MDL Number MFCD08061079
InChI Key OSURJDMYHKQISX-UHFFFAOYSA-N
Synonym 3-3-piperidinyloxy benzonitrile, 3-piperidin-3-yloxy benzonitrile, 3-piperidin-3-yl-oxy benzonitrile, 3-3-piperidyloxy benzenecarbonitrile, 3-piperidin-3-yl oxy benzonitrile
PubChem CID 22309128
IUPAC Name 3-piperidin-3-yloxybenzonitrile
SMILES C1CC(CNC1)OC2=CC=CC(=C2)C#N

Specifications

Quantity 250 mg
Formula Weight 202.26
Percent Purity 99%
Chemical Name or Material 3-(3-Piperidinyloxy)benzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.