missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diethylthiatricarbocyanine iodide, 96%, Thermo Scientific Chemicals

Product Code. 15426927
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3071-70-3
Melting Point 211°C to 212°C
Molecular Formula C25H25IN2S2
MDL Number MFCD00041994,MFCD00041994
Quantity 1 g
UN Number UN2811
Sensitivity Light sensitive
Synonym disc2 7, 3,3'-diethylthiatricarbocyanine iodide, 3-ethyl-2-7-3-ethyl-2 3h-benzothiazolylidene-1,3,5-heptatrienyl benzothiazolium iodide
InChI Key OYVFJKVYVDYPFV-UHFFFAOYSA-M
SMILES [I-].CCN1\C(SC2=CC=CC=C12)=C\C=C\C=CC=CC1=[N+](CC)C2=CC=CC=C2S1
IUPAC Name (2Z)-3-ethyl-2-[7-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)hepta-2,4,6-trienylidene]-1,3-benzothiazole;iodide
Molecular Weight (g/mol) 544.51
PubChem CID 91972095
Formula Weight 544.52
Percent Purity 96%
Chemical Name or Material 3,3'-Diethylthiatricarbocyanine iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.